ENA428683104 Enamine

5-bromo-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD21605341
  • Cas Number : 1328893-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-173989
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart