ENA428683115 Enamine

5-(3-methylpiperazin-1-yl)isoquinoline

  • MDL number MFCD17147804
  • Cas Number : 1483029-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183154
Availability details for
Quantity
  • Availability
  • Add to Cart