ENA428683118 Enamine

(5-nitro-1H-1,3-benzodiazol-2-yl)methanamine

  • MDL number MFCD09038193
  • Cas Number : 115103-10-1
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-174689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-174689
Availability details for
Quantity
  • Availability
  • Add to Cart