ENA428683121 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-5-nitrobenzoic acid

  • MDL number MFCD06220694
  • Cas Number : 777919-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183171
Availability details for
Quantity
  • Availability
  • Add to Cart