ENA428683129 Enamine

4-{[4-(1,3-thiazol-2-yl)piperazin-1-yl]methyl}aniline

  • MDL number MFCD09816721
  • Cas Number : 1016679-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183196
Availability details for
Quantity
  • Availability
  • Add to Cart