ENA428683130 Enamine

5-(2-methylpiperazin-1-yl)isoquinoline

  • MDL number MFCD21452842
  • Cas Number : 1492782-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183208
Availability details for
Quantity
  • Availability
  • Add to Cart