ENA428683138 Enamine

N-(2-aminoethyl)-N-methyloxan-4-amine

  • MDL number MFCD12089073
  • Cas Number : 933737-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183235
Availability details for
Quantity
  • Availability
  • Add to Cart