ENA428683140 Enamine

2-(3-bromo-4-methoxyphenyl)-1H-imidazol-5-ol

  • MDL number MFCD21979966
  • Cas Number : 1368623-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175847
Availability details for
Quantity
  • Availability
  • Add to Cart