ENA428683148 Enamine

N-methylthian-3-amine hydrochloride

  • MDL number MFCD28118424
  • Cas Number : 1645462-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183249
Availability details for
Quantity
  • Availability
  • Add to Cart