ENA428683161 Enamine

N-(5-methylpyridin-2-yl)prop-2-enamide

  • MDL number MFCD12091211
  • Cas Number : 851914-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183288
Availability details for
Quantity
  • Availability
  • Add to Cart