ENA428683167 Enamine

N1-(1-methyl-1H-pyrazol-3-yl)ethane-1,2-diamine

  • MDL number MFCD28118430
  • Cas Number : 1690855-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183298
Availability details for
Quantity
  • Availability
  • Add to Cart