ENA428683172 Enamine

6-chloro-2,3-dihydro-1-benzofuran-3-ol

  • MDL number MFCD23807084
  • Cas Number : 1154740-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183307
Availability details for
Quantity
  • Availability
  • Add to Cart