ENA428683174 Enamine

6-(3-chlorophenyl)pyrimidine-2,4-diamine

  • MDL number MFCD16103651
  • Cas Number : 1251025-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-183310
Availability details for
Quantity
  • Availability
  • Add to Cart