ENA428683175 Enamine

1-butyl-5-ethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD14609663
  • Cas Number : 1251088-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176605
Availability details for
Quantity
  • Availability
  • Add to Cart