ENA428683193 Enamine

thiolane-3-carbonitrile

  • MDL number MFCD19597001
  • Cas Number : 874512-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183349
Availability details for
Quantity
  • Availability
  • Add to Cart