ENA428683205 Enamine

4-(ethylsulfanyl)benzonitrile

  • MDL number MFCD11136354
  • Cas Number : 86047-01-0
  • Molecular Weight : 163.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-183390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183390
Availability details for
Quantity
  • Availability
  • Add to Cart