ENA428683215 Enamine

tert-butyl (2R)-1-(2-hydroxyethyl)pyrrolidine-2-carboxylate

  • MDL number MFCD23463986
  • Cas Number : 1786802-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183404
Availability details for
Quantity
  • Availability
  • Add to Cart