ENA428683222 Enamine

5-chloro-1,3,4-trimethyl-1H-pyrazole

  • MDL number MFCD21667403
  • Cas Number : 1195585-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183429
Availability details for
Quantity
  • Availability
  • Add to Cart