ENA428683227 Enamine

1-[(ethylcarbamoyl)amino]cyclohexane-1-carboxylic acid

  • MDL number MFCD09732010
  • Cas Number : 953752-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183439
Availability details for
Quantity
  • Availability
  • Add to Cart