ENA428683231 Enamine

[4-bromo-2-(trifluoromethyl)phenyl](cyclopropyl)methanone

  • MDL number MFCD20335963
  • Cas Number : 1341736-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183454
Availability details for
Quantity
  • Availability
  • Add to Cart