ENA428683238 Enamine

[1-(4-fluorophenyl)propan-2-yl](methyl)amine hydrochloride

  • MDL number MFCD16294830
  • Cas Number : 52063-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-183677
Availability details for
Quantity
  • Availability
  • Add to Cart