ENA428683251 Enamine

3-(2-chloroethyl)-1-(3-cyanophenyl)urea

  • MDL number MFCD14586696
  • Cas Number : 803729-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183750
Availability details for
Quantity
  • Availability
  • Add to Cart