ENA428683263 Enamine

5-bromo-3-[2-(ethanesulfonyl)ethyl]-3,4-dihydropyrimidin-4-one

  • MDL number MFCD23219492
  • Cas Number : 1538097-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183962
Availability details for
Quantity
  • Availability
  • Add to Cart