ENA428683264 Enamine

6-(trifluoromethyl)pyridine-3,4-diamine

  • MDL number MFCD13181795
  • Cas Number : 438564-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183963
Availability details for
Quantity
  • Availability
  • Add to Cart