ENA428683270 Enamine

4-[3-(trifluoromethyl)phenyl]oxan-4-amine hydrochloride

  • MDL number MFCD22375734
  • Cas Number : 1380300-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183981
Availability details for
Quantity
  • Availability
  • Add to Cart