ENA428683276 Enamine

1-chloro-3-[(1-methoxypropan-2-yl)oxy]propan-2-ol

  • MDL number MFCD26035753
  • Cas Number : 1564601-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184023
Availability details for
Quantity
  • Availability
  • Add to Cart