ENA428683278 Enamine

4-phenylpyrimidin-2-ol

  • MDL number MFCD01646077
  • Cas Number : 38675-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184025
Availability details for
Quantity
  • Availability
  • Add to Cart