ENA428683281 Enamine

1H,2H,3H,4H-pyrrolo[1,2-a]pyrazine

  • MDL number MFCD00296976
  • Cas Number : 71257-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184032
Availability details for
Quantity
  • Availability
  • Add to Cart