ENA428683283 Enamine

1-({5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-3-yl}methyl)-1H-pyrazol-4-amine

  • MDL number MFCD24036073
  • Cas Number : 1521340-43-1
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-184051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184051
Availability details for
Quantity
  • Availability
  • Add to Cart