ENA428683301 Enamine

(2,4,5-trifluorophenyl)thiourea

  • MDL number MFCD17226585
  • Cas Number : 1340519-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-184093
Availability details for
Quantity
  • Availability
  • Add to Cart