ENA428683305 Enamine

2-hydroxy-3-methoxy-4-nitrobenzaldehyde

  • MDL number MFCD25960721
  • Cas Number : 20041-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-94
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184127
Availability details for
Quantity
  • Availability
  • Add to Cart