ENA428683309 Enamine

4,7,8-trichloroquinazoline

  • MDL number MFCD28118473
  • Cas Number : 1027775-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184230
Availability details for
Quantity
  • Availability
  • Add to Cart