ENA428683313 Enamine

1-{4-[2-(5-methylfuran-2-yl)quinoline-4-carbonyl]piperazin-1-yl}ethan-1-one

  • MDL number MFCD10627184
  • Cas Number : 1031195-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184236
Availability details for
Quantity
  • Availability
  • Add to Cart