ENA428683322 Enamine

5-[(1,2,3,4-tetrahydroisoquinolin-2-yl)methyl]quinolin-8-ol

  • MDL number MFCD03968761
  • Cas Number : 22080-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184276
Availability details for
Quantity
  • Availability
  • Add to Cart