ENA428683334 Enamine

ethyl 2,2-difluoro-2-(1-hydroxycyclopentyl)acetate

  • MDL number MFCD12404010
  • Cas Number : 156544-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177794
Availability details for
Quantity
  • Availability
  • Add to Cart