ENA428683337 Enamine

4-amino-6-methyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD00183934
  • Cas Number : 13925-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184367
Availability details for
Quantity
  • Availability
  • Add to Cart