ENA428683343 Enamine

2-ethynyl-3-methylthiophene

  • MDL number MFCD08703515
  • Cas Number : 81294-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184385
Availability details for
Quantity
  • Availability
  • Add to Cart