ENA428683347 Enamine

2-(methoxymethyl)-2-propylpyrrolidine

  • MDL number MFCD21955349
  • Cas Number : 1528597-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184390
Availability details for
Quantity
  • Availability
  • Add to Cart