ENA428683349 Enamine

1-(2-methylpropyl)-5-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13471355
  • Cas Number : 1306678-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178051
Availability details for
Quantity
  • Availability
  • Add to Cart