ENA428683362 Enamine

2-[4-(2-methoxyethoxy)phenyl]acetic acid

  • MDL number MFCD11131696
  • Cas Number : 1019344-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178199
Availability details for
Quantity
  • Availability
  • Add to Cart