ENA428683372 Enamine

7-chloro-1H,2H,3H,4H-pyrazino[1,2-a]indole

  • MDL number MFCD22041113
  • Cas Number : 287384-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184451
Availability details for
Quantity
  • Availability
  • Add to Cart