ENA428683377 Enamine

propane-1-sulfinyl chloride

  • MDL number MFCD28118495
  • Cas Number : 23267-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-184463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184463
Availability details for
Quantity
  • Availability
  • Add to Cart