ENA428683385 Enamine

methyl 1-(2-hydroxyethyl)-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD22371425
  • Cas Number : 65539-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178335
Availability details for
Quantity
  • Availability
  • Add to Cart