ENA428683389 Enamine

5-acetylfuran-2-carboxylic acid

  • MDL number MFCD18431708
  • Cas Number : 13341-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184570
Availability details for
Quantity
  • Availability
  • Add to Cart