ENA428683399 Enamine

[(1S)-1-cyclohexylethyl](methyl)amine

  • MDL number MFCD19217026
  • Cas Number : 138785-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185389
Availability details for
Quantity
  • Availability
  • Add to Cart