ENA428683401 Enamine

ethyl 7-chloro-1H-indole-3-carboxylate

  • MDL number MFCD11617106
  • Cas Number : 1260757-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185391
Availability details for
Quantity
  • Availability
  • Add to Cart