ENA428683403 Enamine

5-bromo-3-(2-methanesulfonylethyl)-6-methyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD23219616
  • Cas Number : 1517442-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185395
Availability details for
Quantity
  • Availability
  • Add to Cart