ENA428683472 Enamine

[2-(ethylamino)ethyl](methyl)(prop-2-en-1-yl)amine dihydrochloride

  • MDL number MFCD28118508
  • Cas Number : 1795281-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185437
Availability details for
Quantity
  • Availability
  • Add to Cart