ENA428683475 Enamine

5-[(2,6-difluorophenyl)methyl]-4-methyl-1,3-thiazol-2-amine hydrobromide

  • MDL number MFCD28118511
  • Cas Number : 1795486-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185445
Availability details for
Quantity
  • Availability
  • Add to Cart