ENA428683479 Enamine

3-(3-chloropropyl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD28118520
  • Cas Number : 1803593-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185536
Availability details for
Quantity
  • Availability
  • Add to Cart